Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CGS 21680 hydrochloride, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
CGS 21680 hydrochloride is a potent A2A adenosine receptor agonist (Ki = 27 nM, EC50 = 1.48-180 nM). CGS 21680 reduces venous resistance, increases heart rate and cardiac output in vivo. In an animal model of Huntington's disease, CGS21680 slows motor deterioration, prevents reduction in brain weight and reduces the size of mutant huntingtin aggregates and intranuclear inclusions. CGS21680 increases proliferation of neural progenitor cells, promotes survival of motoneurons and activates TrkB receptors. Also has anti-inflammatory effects in animal models of lung injury and reduces neurotoxicity induced by Kainate (Cat. No. 7065).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 535.99 |
Percent Purity | >98% |
Chemical Name or Material | CGS 21680 hydrochloride |
Safety and Handling
Recommended Storage | Desiccate at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction