Learn More
Teknova CGXII Minimal Media. 1000mL, Sterile. 3/UN1993/II (L) - 5L limited 2 pack.
SDP
Supplier:  Teknova C89152PK
Encompass
2.000% Ammonium Sulfate,0.500% Urea,0.100% Potassium Phosphate Monobasic,0.100% Potassium Phoshpate Dibasic,0.025% Magnesium Sulfate,4.200% MOPS,68.00µM Calcium Chloride,36.00µM Iron Sulfate,10.000mg/L Manganese Sulfate,3.480µM Zinc Sulfate,0.200mg/L Copper Sulfate,0.084µM Nickel Chloride,4.000% Glucose,100mM Protocatechuic Acid
Catalog No. 50-238-7356
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.