Learn More
Biotium Cheetah™ Hot Start Taq DNA Polymerase
Supplier: Biotium 29050
Cheetah™ Taq is a chemically modified hot-start Taq DNA polymerase useful for preventing or reducing nonspecific DNA amplification in PCR. Modified using a novel modifying reagent, Cheetah™ Taq represents a major improvement upon AmpliTaq Gold by having a faster activation time and better shelf-life. Under standard hot-start conditions (i.e., 94°C in pH 8-9 Tris), Cheetah™ Taq regains both the 3'-5' polymerase and 5'-exonuclease activities within two minutes. Cheetah™ Taq is also superior to antibody-based hot-start DNA polymerases due to its complete supression of enzyme activity at room temperature and full recovery of enzyme activity following activation. Cheetah™ Taq is covered by patent US patents. AmpliTaq is a registered trademark of Thermo Fisher Scientific.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.