Learn More
MilliporeSigma™ Chemicon™ Fibronectin, Bovine
Shop All MilliporeSigma ProductsSpecifications
Concentration
1 mg/mL
For Use With (Application)
Cell Culture
Formulation
Liquid in a solution of 2M Urea in 0.05 M Tris pH 7.0.
pH Range
7
Quantity
500 μg
Source
Bovine Plasma
Product Type
Fibronectin, Bovine
Purity or Quality Grade
95% by SDS-PAGE
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.