Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Mouse Spinal Cord Neural Stem Cell Expansion Kit

Catalog No. SCR033 Shop All MilliporeSigma Products
Click to view available options
:

Used in Immunocytochemistry, Stem cell culture applications. MilliporeSigma™; Chemicon™ Mouse Spinal Cord Neural Stem Cell Expansion Kit provides a multi-component system for the culture and analysis of neural stem cells and their differentiated progenies.

  • Includes primary neural stem cells isolated from the spinal cord of embryonic day 15-18 (E15-E18) C57/BL6 mice, neural stem cell expansion medium, and a panel of antibodies for immunocytochemical staining of neural stem/progenitor cells (Nestin and Sox2) and differentiated neural phenotypes (Map2ab for neurons, GFAP for astrocytes, and O1 for oligodendrocytes).

Specifications

Content And Storage

  • Cryopreserved Mouse Spinal Cord Neural Stem Cells (Cat.# SCR031)
  • Mouse Neural Stem Cell Expansion Medium (Cat.# SCM008)
  • Neural Stem Cell Marker Characterization Kit (Cat.# SCR019)

Cell Type

Neural Stem Cells

Product Type

Stem Cell Expansion Media

For Use With (Application)

Immunocytochemistry, Stem cell culture

Quantity

1 Kit

Regulatory Status

RUO

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.