Learn More
TARGETMOL CHEMICALS INC CHIKUSETSUSAPONIN IVA 5MG
Supplier:  TARGETMOL CHEMICALS INC T3863
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Chikusetsusaponin IVa (Calenduloside F) is a novel AMPK activator can induce insulin secretion from (beta)TC3 cells via GPR4 mediated calcium and PKC pathways may be developed into a new potential for therapeutic agent used in T2DM patients. purity: 99%
Catalog No. 50-258-0097
missing translation for 'QuantityLeft'