Learn More
Ergodyne™ Chill-Its™ 6235 Standard Phase Change Cooling Vest
Shop All Ergodyne Products- Adjustable hook and loop closures on both shoulders and waist
- Constructed of cotton
- Extended vest surface area enhances cooling power
Specifications
Material
Cotton
Product Type
Cooling Vest
Size
Small/Medium
Length (Metric)
81 to 102 cm
Length (English)
32 to 40 in.
Color
Tan
For Use With (Application)
Fire, Rescue, EMT, Construction, Utilities
Quantity
6/Pk.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.