Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHIR 99021, Tocris Bioscience™
GSA_VA

Catalog No. p-4816967 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Highly selective GSK-3 inhibitor

CHIR 99021 is a potent and highly selective inhibitor of glycogen synthase kinase 3 (GSK-3) (IC50 values are 6.7 and 10 nM for GSK-3β and GSK-3α respectively). Acts as Wnt activator. Exhibits >500-fold selectivity for GSK-3 over closely related kinases; also displays >800-fold selectivity against 45 additional enzymes and receptors. In combination with the irreversible inhibitor of LSD1 (Cat. No. 3852), CHIR 99021 enables reprogramming of mouse embryonic fibroblasts, transduced by Oct4 and Klf4 only, into iPSCs. Enhances mouse and human ESC self-renewal when used in combination with PD 0325901 (Cat. No. 4192). It promotes differentiation of hPSCs in cardiomyocytes and allows formation of extended pluripotent stem (EPS) cells in combination with (S)-(+)-Dimethindene maleate (Cat. No. 1425), tetracycline antibiotic (Cat. No. 3268) and human leukemia inhibitory factor. CHIR 99021 synthesized to cGMP guidelines also available. For more information on how to use CHIR 99021, see our protocols: Transdifferentiating Fibroblasts into Neurons, Culturing Transcriptomically Distinct Pluripotent mESCs, Converting Fibroblasts into Cardiomyocytes (9C Cocktail), Human iPSC-derived Myoblasts .

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 252917-06-9
Molecular Formula C22H18Cl2N8
Molecular Weight (g/mol) 465.34
MDL Number MFCD11846251
InChI Key AQGNHMOJWBZFQQ-UHFFFAOYSA-N
Synonym gsk-3 inhibitor xvi, unii-234cmt4gk4, chir-911, 6-2-4-2,4-dichlorophenyl-5-4-methyl-1h-imidazol-2-yl pyrimidin-2-ylamino ethylamino pyridine-3-carbonitrile, 6-2-4-2,4-dichlorophenyl-5-4-methyl-1h-imidazol-2-yl pyrimidin-2-yl amino ethyl amino nicotinonitrile, 6-2-4-2,4-dichlorophenyl-5-4-methyl-1h-imidazol-2-yl pyrimidin-2-yl amino ethyl amino pyridine-3-carbonitrile, 6-2-4-2,4-dichlorophenyl-5-5-methyl-1h-imidazol-2-yl-2-pyrimidinyl amino ethyl amino-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 6-2-4-2,4-dichlorophenyl-5-5-methyl-1h-imidazol-2-yl-2-pyrimidinyl amino ethyl amino, 6-2-4-2,4-dichlorophenyl-5-5-methyl-1h-imidazol-2-yl pyrimidin-2-yl amino ethyl amino nicotinonitrile
PubChem CID 9956119
IUPAC Name 6-[(2-{[4-(2,4-dichlorophenyl)-5-(5-methyl-1H-imidazol-2-yl)pyrimidin-2-yl]amino}ethyl)amino]pyridine-3-carbonitrile
SMILES CC1=CN=C(N1)C1=CN=C(NCCNC2=NC=C(C=C2)C#N)N=C1C1=C(Cl)C=C(Cl)C=C1

Specifications

Quantity 10 mg
Formula Weight 465.34
Percent Purity >99%
Chemical Name or Material CHIR 99021
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.