Learn More
Strem, An Ascensus Company CAS 791616-63-2. 100mg. (11bR)-4-Hydroxy-26-bis[246-tris(1-methylethyl)phenyl]-4-oxide-dinaphtho[21-d12-f][132]dioxaphosphepin 98% (99% ee).
SDP
Supplier:  Strem, An Ascensus Company 151381100MG
Encompass_Preferred
CAS 791616-63-2. 100mg. (11bR)-4-Hydroxy-26-bis[246-tris(1-methylethyl)phenyl]-4-oxide-dinaphtho[21-d12-f][132]dioxaphosphepin 98% (99% ee). . Molecular Weight 753.0. Molecular Formula C50H57O4P. Color/form white to light-yellow pwdr. Strem 15-1381. http//www.strem.com/catalog/v/15-1381/
Catalog No. 50-211-0747
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.