Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorhexidine Digluconate Aqueous Solution 0.2 MP Biomedicals

Catalog No. p-3592997 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
25 mL
500 mL

Chemical Identifiers

CAS 18472-51-0
Molecular Formula C34H54Cl2N10O14
Molecular Weight (g/mol) 897.76
MDL Number MFCD00083599
InChI Key YZIYKJHYYHPJIB-UUPCJSQJSA-N
Synonym peridex, periogard, chlorhexidine gluconate, 1,1'-hexamethylene bis 5-p-chlorophenyl biguanide di-d-gluconate
PubChem CID 60148207
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1

Specifications

Quantity 25 mL
IUPAC Name bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid); N'-(6-{[amino({amino[(4-chlorophenyl)amino]methylidene}amino)methylidene]amino}hexyl)-1-({amino[(4-chlorophenyl)amino]methylidene}amino)methanimidamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.