Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloro(ethyl)dimethylsilane 97.0+%, TCI America™
SDP

Catalog No. p-7126054 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 6917-76-6
Molecular Formula C4H11ClSi
Molecular Weight (g/mol) 122.667
MDL Number MFCD00009918
InChI Key AVDUEHWPPXIAEB-UHFFFAOYSA-N
Synonym ethyldimethylchlorosilane, chlorodimethylethylsilane, chloro ethyl dimethylsilane, dimethylethylchlorosilane, dimethylethylsilyl chloride, ethyldimethylsilyl chloride, chloro ethyl dimethylsilane #, silane, chlorodimethylethyl-,
PubChem CID 138863
IUPAC Name chloro-ethyl-dimethylsilane
SMILES CC[Si](C)(C)Cl

Specifications

Color Yellow
Boiling Point 88°C
Quantity 5 g
UN Number 2985
Formula Weight 122.67
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloro(ethyl)dimethylsilane [Dimethylethylsilylating Agent]
EINECSNumber (2)-2041
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.