Learn More
Chlorobis(2-methoxyphenyl)phosphine, 98+%, Thermo Scientific Chemicals
Description
Chlorobis(2-methoxyphenyl)phosphine is used as a reactant for cycloadditions reactions, asymmetric hydrogenation and allylic substitution, asymmetric hydrogenation of imines and olefins. It is also used as a reactant for synthesis of ferrocene-based chiral diphosphine, catalysts for C-C cross coupling reactions, palladium complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chlorobis(2-methoxyphenyl)phosphine is used as a reactant for cycloadditions reactions, asymmetric hydrogenation and allylic substitution, asymmetric hydrogenation of imines and olefins. It is also used as a reactant for synthesis of ferrocene-based chiral diphosphine, catalysts for C-C cross coupling reactions, palladium complexes.
Solubility
Reacts with water.
Notes
Air and moisture sensitive. Incompatible with oxidizing agents, bases, water/ moisture.
Specifications
Specifications
Melting Point | 111°C to 114°C |
Quantity | 1 g |
UN Number | UN3261 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Reacts with water. |
Formula Weight | 280.69 |
Percent Purity | ≥98% |
Chemical Name or Material | Chlorobis(2-methoxyphenyl)phosphine |
Safety and Handling
Hazard Category | H314-H335 |
Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
EINECSNumber | 000-000-0 |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.