Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Chloromethyl)trimethoxysilane 98.0+%, TCI America™
SDP

Catalog No. p-7125811 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 5926-26-1
Molecular Formula C4H11ClO3Si
Molecular Weight (g/mol) 170.66
MDL Number MFCD00468107
InChI Key FPOSCXQHGOVVPD-UHFFFAOYSA-N
PubChem CID 22212
IUPAC Name (chloromethyl)trimethoxysilane
SMILES CO[Si](CCl)(OC)OC

Specifications

Color Colorless
Boiling Point 156°C
Quantity 5 g
UN Number 1993
Formula Weight 170.66
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (Chloromethyl)trimethoxysilane
EINECSNumber (2)-2079
RTECSNumber VV2625000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.