Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloronitrosyl[N,N′-bis(3,5-di-tert-butylsalicylidene)-1,1,2,2-tetramethylethylenediaminato]ruthenium(IV), TCI America™
SDP

Catalog No. C1944100MG Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg

Chemical Identifiers

CAS 386761-71-3
Molecular Formula C36H56ClN3O3Ru
Molecular Weight (g/mol) 715.38
MDL Number MFCD06797082
InChI Key ZQSCHIZFMGABNG-UHFFFAOYSA-M
PubChem CID 121233811
IUPAC Name 2,4-di-tert-butyl-6-{[(3-{[(3,5-di-tert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}-2,3-dimethylbutan-2-yl)amino]methylidene}cyclohexa-2,4-dien-1-one; chlororutheniumylium; oxoazanide
SMILES Cl[Ru+].[N-]=O.CC(C)(C)C1=CC(=CNC(C)(C)C(C)(C)NC=C2C=C(C=C(C2=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C

Specifications

Quantity 100 mg
Formula Weight 713.37
Physical Form Crystalline Powder
Chemical Name or Material Chloronitrosyl[N,N′-bis(3,5-di-tert-butylsalicylidene)-1,1,2,2-tetramethylethylenediaminato]ru
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.