Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorothiazide, 98%, Thermo Scientific Chemicals

Catalog No. p-7112891
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 58-94-6 | C7H6ClN3O4S2 | 295.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58-94-6
Molecular Formula C7H6ClN3O4S2
Molecular Weight (g/mol) 295.71
MDL Number MFCD00058576
InChI Key JBMKAUGHUNFTOL-UHFFFAOYSA-N
Synonym chlorothiazide, diuril, chlorthiazide, chlorothiazid, chlotride, chlortiazid, thiazide, flumen, chlorosal, chlorurit
PubChem CID 2720
ChEBI CHEBI:3640
IUPAC Name 6-chloro-1,1-dioxo-4H-1$l^{6},2,4-benzothiadiazine-7-sulfonamide
SMILES NS(=O)(=O)C1=CC2=C(NC=NS2(=O)=O)C=C1Cl

Specifications

Melting Point 348.0°C
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Solubility Information Solubility in water: very slightly soluble. Other solubilities: freely soluble in dmso, dmf, slightly soluble in methanol, pyridine, practically insoluble in diethyl ether, benzene, and chloroform
Formula Weight 295.72
Percent Purity 98%
Chemical Name or Material Chlorothiazide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
In case of inadequate ventilation wear respiratory protecti
EINECSNumber 200-404-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.