Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorpropham 99.0+%, TCI America™
SDP

Catalog No. p-7125526 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
25 g

Chemical Identifiers

CAS 101-21-3
Molecular Formula C10H12ClNO2
Molecular Weight (g/mol) 213.66
MDL Number MFCD00037108
InChI Key CWJSHJJYOPWUGX-UHFFFAOYSA-N
Synonym chlorpropham, chloropropham, chloro-ipc, isopropyl m-chlorocarbanilate, cipc, chlorprophame, elbanil, isopropyl n-3-chlorophenyl carbamate, metoxon, nexoval
PubChem CID 2728
ChEBI CHEBI:34630
IUPAC Name propan-2-yl N-(3-chlorophenyl)carbamate
SMILES CC(C)OC(=O)NC1=CC=CC(Cl)=C1

Specifications

Melting Point 42°C
Color White-Yellow
Quantity 1 g
UN Number 3077
Formula Weight 213.66
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Chlorpropham
EINECSNumber (3)-3217
RTECSNumber FD8050000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.