Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ ChromatoPur™ Bovine Thrombin, Low Specific Activity, Diagnostic Grade

Catalog No. p-4341501 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
10 kU
100 kU
2 kU
25 kU

Suitable as a diagnostic and research reagent in clotting assays

Chromatographically purified with minimal loss of native clotting activity and minimized degradation to the gamma form. Enzyme activity is measured by clotting assay. 90 to 300U/mg protein.

Physical Characteristics

  • Appearance: White to off-white powder
  • pH: 7.0 to 8.0
  • Protein: ≥50%
  • Specific activity: 90 to 300U/mg protein
  • Activity: Report μ/mg solid
  • Total Activity: U/vial
  • Exipients: Reported
Certifications: ISO, cGMP

Specifications

For Use With (Application) Cell Culture, Microbial Culture
Name Bovine Thrombin
pH Range 7 to 8
Quantity 100 kU
Source Bovine Thrombin
Storage Requirements -10°C
Form Solid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.