Learn More
Andwin Scientific CHROMOLITH SPEEDROD RP-18
SDP
Supplier:  Andwin Scientific 1.51450.0001
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
The Merck Chromolith HPLC columns is ideal for use in combinatorial chemistry. The columns offer a quick and high-quality performance of separating acidic, basic, nonpolar, and metal chelating compounds. This product has a dimension of 4.6 x 50 mm, a macropore size of 2 mm, a micropore size of 13 nm, a pore volume of 1 mL/g, and a surface area of 300 square meters/g.
Catalog No. NC9849834