Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromplete™ 2-Propanol, HPLC/GC, meets ACS/USP analytical requirements, Thermo Scientific™

Catalog No. p-8776010
Click to view available options
Quantity:
1 L
4 L
25 L

The Chromplete product line from Thermo Scientific includes reagents for common analyses and separations with the consistency and precision you expect. Chromplete Isopropanol is suitable for HPLC, GC, SFC, TLC, ACS, and Spectrophotometry.

Chromplete Isopropanol meets the analytical requirements of USP and is suitable for use in:

  • HPLC coupled with UV, VIS, PDA, ECD, FL, and RI detectors
  • GC- FID, GC- ECD, GC-TCD
  • Pesticide Analysis by GC or HPLC
  • UV Spectrophotometry
  • Preparative chromatography
  • Supercritical Fluid and Thin Layer chromatography
  • Reactions/Analyses requiring ACS compliance
  • Extraction/Concentration Analyses

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -90°C
Absorbance ≤ 1.0, ≤ 0.20, ≤ 0.10, ≤ 0.015
Color ≤ 5 APHA
CAS Min % 0.999
Boiling Point 83°C
Quantity 1 L
Packaging Amber Glass
Physical Form Liquid
Residue after Evaporation ≤ 1.0 ppm
UN Number 1219
Water ≤0.05%
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
Molecular Weight (g/mol) 60.1
Formula Weight 60.1
Percent Purity 99.9%
Grade Chromplete
Assay Minimum, GC-FID
Chemical Name or Material 2-Propanol
Show More Show Less
Hazard Category Serious eye damage/eye irritation (category 2), Flammable liquids (category 2), Specific target organ toxicity after single exposure (category 3)
Hazard Statement Causes serious eye irritation, Highly flammable liquid and vapor, May cause drowsiness or dizziness
ShelfLife 1825 days
EINECSNumber 200-661-7
RTECSNumber NT8050000
storageNote1 At room temperature in sealed containers
Recommended Storage Room temperature flammable storage

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.