Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CID 2745687, Tocris Bioscience™

Catalog No. p-4816982 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

GPR35 antagonist

CID 2745687 is a competitive, reversible antagonist of the orphan receptor GPR35 (Ki = 12.8 nM). Blocks the GPR35-mediated increase in ERK1/2 phosphorylation and β-arrestin recruitment induced by pamoic acid.

Chemical Identifiers

CAS 264233-05-8
Molecular Formula C17H19F2N5O2S
Molecular Weight (g/mol) 395.429
InChI Key CYNLZIBKERMMOA-AWQFTUOYSA-N
Synonym 1-2,4-difluorophenyl-5-2-1,1-dimethylehyl amino thioxomethyl hydrazinylidene methyl-1h-pyrazole-4-carboxylic acid methyl ester, methyl 5-2-tert-butylamino carbothioyl carbohydrazonoyl-1-2,4-difluorophenyl-1h-pyrazole-4-carboxylate, ml hplc, 5-e-tert-butylthiocarbamoylhydrazono methyl-1-2,4-difluorophenyl pyrazole-4-carboxylic acid methyl ester, 5-tert-butylthiocarbamoylhydrazono methyl-1-2,4-difluorophenyl pyrazole-4-carboxylic acid methyl ester, 5-tert-butylamino-sulfanylidenemethyl hydrazinylidene methyl-1-2,4-difluorophenyl-4-pyrazolecarboxylic acid methyl ester, methyl 1-2,4-difluoro-phenyl-5-4-tert-butyl-thiosemicarbazono methyl-1h-pyrazole-4-carboxylate, methyl 1-2,4-bis fluoranyl phenyl-5-tert-butylcarbamothioylhydrazinylidene methyl pyrazole-4-carboxylate
PubChem CID 9581011
IUPAC Name methyl 5-[(E)-(tert-butylcarbamothioylhydrazinylidene)methyl]-1-(2,4-difluorophenyl)pyrazole-4-carboxylate
SMILES CC(C)(C)NC(=S)NN=CC1=C(C=NN1C2=C(C=C(C=C2)F)F)C(=O)OC

Specifications

Quantity 10 mg
Formula Weight 395.43
Percent Purity >98%
Chemical Name or Material CID 2745687
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.