Learn More
Selleck Chemical LLC Cilostazol 100mg 73963-72-1 OPC-13013
Supplier:  Selleck Chemical LLC S1294100MG
Encompass_Preferred
Cilostazol (OPC-13013) is a potent cyclic nucleotide phosphodiesterase type 3 (PDE3) inhibitor with IC50 of 0.2 ?M and inhibitor of adenosine uptake. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-764-1
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.