Learn More
Medchemexpress LLC HY-66010 50mg Medchemexpress, Cinepazide (Maleate) CAS:26328-04-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY6601050MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-66010 50mg Cinepazide (Maleate) CAS:26328-04-1 Cinepazide Maleate (MD-67350) is a piperazine derivative and acts as a weak calcium channel blocker. Cinepazide Maleate is a potent vasodilator and can be used for the research of cerebrovascular diseases, including ischemic stroke, brain infarct et. al.
Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2265