Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cinnamic Aldehyde, 99%, MP Biomedicals

Catalog No. p-3547072 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 g
500 g

Chemical Identifiers

CAS 104-55-2
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.162
InChI Key KJPRLNWUNMBNBZ-QPJJXVBHSA-N
Synonym cinnamaldehyde, trans-cinnamaldehyde, cinnamic aldehyde, e-cinnamaldehyde, cinnamal, 3-phenylacrylaldehyde, zimtaldehyde, cinnamylaldehyde, 3-phenylpropenal, 2e-3-phenylprop-2-enal
PubChem CID 637511
ChEBI CHEBI:16731
IUPAC Name (E)-3-phenylprop-2-enal
SMILES C1=CC=C(C=C1)C=CC=O

Specifications

Assay Percent Range 99%
Quantity 100 g
Formula Weight 132.2
Concentration or Composition (by Analyte or Components) 1mL = approx. 1.05g
Chemical Name or Material Cinnamic Aldehyde
Recommended Storage Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.