Learn More
Cole-Parmer Techne FBATH48 Unheated Water Bath, Stainless Steel, 48L
Supplier: Cole-Parmer 1610445
Designed to be used with a clip-on Tempette or Tempunit thermoregulator (sold separately), these water baths incorporate carrying handles for added safety. All water baths have stoved enamelled steel outer cases and are supplied with bridge mounting plate to hold the thermoregulator.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.