Learn More
Cole-Parmer Techne TU-20D Advanced Digital Immersion Circulator; 230 V, 50/60 Hz
Supplier:  Cole-Parmer 0126215
Encompass_Preferred
This digital immersion circulator offers a wide temperature range of 40 to 200°C (with optional chiller). Circulator includes all of the features found on the basic digital circulator model, plus RS-232 interface capability. Bidirectional RS-232 lets you control bath remotely and download data to a computer for further analysis.
Catalog No. 50-196-0467
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.