Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic Acid 97.0+%, TCI America™
SDP

Catalog No. p-7122998 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 222530-33-8
Molecular Formula C12H21NO4
Molecular Weight (g/mol) 243.303
MDL Number MFCD03453252
InChI Key JSGHMGKJNZTKGF-BDAKNGLRSA-N
Synonym 1r,3s-3-tert-butoxycarbonylamino cyclohexanecarboxylic acid, 1r,3s-3-boc-amino-cyclohexanecarboxylic acid, cis-+/--3-boc-amino cyclohexanecarboxylic acid, boc-cis-3-aminocyclohexanecarboxylic acid, cis-3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,3s-rel-3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,3s-3-boc-amino cyclohexanecarboxylic acid, cis-3-boc-amino cyclohexanecarboxylic acid, cis-3-tert-butoxycarbonylamino cyclohexanecarboxylic acid, 1r,3s-3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid
PubChem CID 16218560
IUPAC Name (1R,3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1CCCC(C1)C(=O)O

Specifications

Melting Point 145°C
Color White-Yellow
Quantity 1 g
Formula Weight 243.30
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material cis-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.