Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Citalopram hydrobromide

Catalog No. p-7112908
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59729-32-7
Molecular Formula C20H21FN2O·HBr
Molecular Weight (g/mol) 405.3
InChI Key WIHMBLDNRMIGDW-UHFFFAOYSA-N
Synonym citalopram hydrobromide, citalopram hbr, cipramil, celexa, 1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydroisobenzofuran-5-carbonitrile hydrobromide, elopram, seropram, 1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydro-5-isobenzofurancarbonitrile hydrobromide, citalopram hydrobromide usan
PubChem CID 77995
IUPAC Name 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile;hydrobromide
SMILES CN(C)CCCC1(C2=C(CO1)C=C(C=C2)C#N)C3=CC=C(C=C3)F.Br

Specifications

Melting Point 185.0°C to 188.0°C
Quantity 1 g
Solubility Information (1% in ethanol) Clear colorless
Formula Weight 405.3
Chemical Name or Material Citalopram hydrobromide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face, hands and any exposed skin thoroughly after handling.
EINECSNumber 261-890-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.