Learn More
Citric Acid, Monohydrate, USP, EP, BP, JP, FCC, bioCERTIFIED™, 1 kg, Spectrum Chemical
SDP
Catalog No. 18615339 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
:
Quantity: 1 kg
Citric Acid, Monohydrate, USP, EP, BP, JP, FCC, bioCERTIFIED &rade; is used as an acidulant to control pH and to passivate high purity process piping. All Spectrum Chemical USP, EP, BP, JP, FCC bioCERTIFIED™ grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Specifications
Quantity 1 kg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.