Learn More
Citric Acid, Monohydrate, USP, EP, BP, JP, FCC, bioCERTIFIED™, 50 kg, Spectrum Chemical
SDP
Catalog No. 18615589 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
:
Quantity: 50 kg
Citric Acid, Monohydrate, USP, EP, BP, JP, FCC, bioCERTIFIED &rade; is used as an acidulant to control pH and to passivate high purity process piping. All Spectrum Chemical USP, EP, BP, JP, FCC bioCERTIFIED™ grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Specifications
Quantity 50 kg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.