Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Cladribine
Deoxyadenosine analog; pro-apoptotic
Specifications
Purity | >99% |
---|---|
Product Type | Cladribine |
Molecular Weight (g/mol) | 285.69 |
Inhibitors | Cladribine |
Description
Adenosine deaminase (ADA)-resistant analog of deoxyadenosine. Exhibits anti-leukemic activity; Inhibits cell growth of lymphoid and myeloid neoplasm (IC50 = 20 - 87 nM). Pro-apoptotic; induces DNA double-strand breaks. Also inhibits ribonuclease reductase (IC50 = 0.11 - 0.28 μM). Displays activity in vitro and in vivo. Orally bioavailable.Specifications
>99% | |
Cladribine | |
285.69 | |
Cladribine |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title