Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ CleanTeam™ Premium Lightweight Cotton Lisle Inspection Gloves with Overcast Hem

Catalog No. p-7230591 Shop All Protective Industrial Products Inc. Products
Click to view available options
Gender:
Men's
Women's
This item is not returnable. View return policy
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
This item is not returnable. View return policy
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit

Gloves are constructed in a two-piece, reversible pattern using 100% cotton fabric. Wear alone for use with inspection and electronics assembly, or as glove liners under disposable and unsupported liquid-proof gloves.

  • Constructed in a two piece, reversible pattern using 100% cotton fabric
  • Designed to minimize contamination of products from human hands
  • Can be used as liners under disposable and unsupported liquid proof gloves
  • Cut-and-Sewn construction
  • Overcast hem

Specifications

Construction Cut and Sewn
Coating Material Uncoated
Color White
Grade Premium
Material Cotton
Product Type Inspection Gloves
Style Reversible
For Use With (Application) Electronics Assembly, Inspection
Gender Men's
Glove Cuff Style Overcast Hem
Series 97-500
Length (English) 8.9 in.
Length (Metric) 22.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.