Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ CleanTeam™ Seamless Knit Nylon/Carbon Fiber ESD Gloves with PVC Dot Grip
GSA_VA

Catalog No. p-7230589 Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
Small
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Electrostatic Dissipative (ESD) fiber yarns blended with low lint nylon reduce static build-up on the glove's surface for improved performance in electronics assembly. PVC dots enhance grip capabilities, and provide additional cut/abrasion resistance.

  • Electrostatic Dissipative (ESD) fiber yarns blended with low lint nylon
  • Reduced static build-up on the glove's surface for improved performance in electronics assembly
  • PVC dots on the palm enhance grip capabilites, and provide additional cut/abrasion resistance
  • Electrical Surface Resistivity (ASTM D 257:2007): Below 7.5 x 107
  • Seamless knit construction enhances dexterity
  • Launderable without losing ESD properties
  • Reusable

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Coated Area Palm
Construction Seamless Knit
Coating Material PVC Dots
Color Gray
Disposable Reusable
Hand Specific
Material Nylon, Carbon Fiber
Product Type ESD Gloves
Size Small
For Use With (Application) Aerospace, Circuit Board Manufacturing, Electronics Manufacturing, and Munitions
Glove Cuff Style Continuous Knit
Hem Color Orange
Palm Type Coated
Series 40-6411
Yarn Gauge 15
Length (English) 7.9 in.
Length (Metric) 20 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.