Learn More
Cleatech PP 32" VER LAMINAR FLOW HOOD
SDP
Supplier:  Cleatech 10007AFL
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Portable Bench top Vertical Laminar Flow station, Work space 31.5"Wx 21.75"Dx 23.75"H , Polypropylene construction, Clear Acrylic front tilt-up sash,110VAC, 50/60Hz. Adjustable speed Impeller, Backward Curve, 730 CFM @ 0.000-In,Including HEPA Filter , Including Built-in Fluorescent Light
Catalog No. 50-152-7910
missing translation for 'QuantityLeft'