Learn More
Electron Microscopy Sciences Micro Cleveland Bone Rongeur, 4 mm
Supplier:  Electron Microscopy Sciences 7798604
Encompass_Preferred
Bone Rongeur with curved jaws. Made from stainless steel with a polished finish. Curved jaws, with a 4mm jaw width, 5.5" long.
Catalog No. 50-301-81
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.