Learn More
Lakeland Industries CLEANMAX BULK PACK NonSterile Microporous Cleanroom coverall. Bound seams, elastic back, wrists, ankles, thumbloops, zipper placket. 25 per case with overlay bag. Size 3X
Supplier: Lakeland Industries CTL417CMB3X
CleanMax Microporous Cleanroom BULK Apparel is available in non-sterile configurations only. Clean-Manufactured under strict process and environment controls in Lakeland owned, operated, and certified cleanrooms. CleanMax cleanroom garments meet IEST Cleanliness levels. 99.999% bacterial and particulate filtration makes CleanMax resistant to microbial, viral and blood borne contaminants, along w/ most API’s, HD’s, and some cytotoxic drugs. Easy donning fold, zipper plackets, elastic back in the coveralls, thumb loops and always bound seams provide a superior barrier to prevent breakthrough and strike through. Let Lakeland clean-manufactured CleanMax apparel be the cornerstone of your contamination control strategy. Coveralls are constructed w/ a fluid-repellant, low-linting, microporous fabric. Bound neckline, zipper w/ placket, elastic wrist w/ thumb loops, elastic ankles and back for a more tailored fit. BULK PACKAGED IN OVERLAY BAG, 25/case.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.