Learn More
Bevco Precision Manufacturing Co Doral Mid Height ISO 5 Cleanroom Black Vinyl Chair; Articulating Seat & Back Tilt; Chrome Tubular Steel Base w/Welded Footring
Supplier: Bevco Precision Manufacturing Co 5611C2BLK
BEVCO’s Doral ISO 5 Cleanroom Series models are designed to meet the particulate goals of ISO 14644-1 . A unique filtration system traps particles for contamination-free comfort has been tested by an independent laboratory to certify ISO 5 compliance. This articulating tilt model features simultaneous seat and back adjustments to properly position the angle of the chair for increased comfort during extended sitting durations. Fully adjustable production-style back has a durable scuff resistant back pan to protect the upholstery. A waterfall seat promotes lower body circulation, improving overall wellness. The pneumatic height adjustment allows you to easily change your seat height from 25" - 30". Black vinyl meets CAL 117 standards. The chair features a sturdy 22" diameter chrome tubular steel base with welded footring. Model comes standard with mushroom glides. Optional casters can be ordered separately.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.