Learn More
Selleck Chemical LLC Clozapine N-oxide CNO-5mg
Supplier:  Selleck Chemical LLC S68875MG
Encompass_Preferred
Clozapine N-oxide (CNO) is a metabolite of Clozapine (GLXC-06516) and an agonist of human muscarinic designer receptors (Designer Receptors Exclusively Activated by Designer Drugs, DREADDs). Clozapine is a potent dopamine (DA) antagonist and a selective muscarinic M4 receptor agonist.Solutions are unstable and should be fresh-prepared.
Catalog No. 50-238-9948
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.