Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clozapine N-oxide, Tocris Bioscience™

Catalog No. p-4816900 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Activator of muscarinic DREADDs

Metabolite of clozapine (Cat. No. 0444). Activates human muscarinic designer receptors (DREADDs) in vitro. Silences hippocampal neurons expressing hM4 DREADDs in vitro. Activates Gs-linked DREADDS- AgRP in mice via intracerebroventricular (i.c.v) injection. Shown to be a P-glycoprotein (P-gp) efflux pump substrate. Other potent and selective hM3Dq DREADD agonists available: DREADD agonist 21 and Perlapine .

Water soluble salt also available.

This product contains 1 molar equivalent of ethanol.

Chemical Identifiers

CAS 34233-69-7
Molecular Formula C18H19ClN4O
Molecular Weight (g/mol) 342.827
InChI Key WYRDWWAASBTJLM-UHFFFAOYSA-N
Synonym clozapine n-oxide, clozapine-n-oxide, unii-mza8bk588j, chembl1688, mza8bk588j, n-oxyclozapine, clozapine n-oxide cno, oguczbiqsyywef-uhfffaoysa-n
PubChem CID 2819
IUPAC Name 3-chloro-6-(4-methyl-4-oxidopiperazin-4-ium-1-yl)-5H-benzo[b][1,4]benzodiazepine
SMILES C[N+]1(CCN(CC1)C2=C3C=CC=CC3=NC4=C(N2)C=C(C=C4)Cl)[O-]

Specifications

Assay Percent Range >98%
Quantity 10 mg
Formula Weight 342.82
Percent Purity >99%
Chemical Name or Material Clozapine N-oxide
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.