Learn More
Electron Microscopy Sciences CMCP-10 High Viscosity Mountant 250 ML
Supplier:  Electron Microscopy Sciences 1800402
Encompass_Preferred
Colorless, non resinous, water miscible mounting medium is used like Canada Balsam to mount almost any material, provided it is not stained, calciferous material, or thin tissue sections. This permanent transparent mounting media has the advantage of allowing live or preserved specimens to be mounted directly from water or alcohol. Free living nematodes and live parasitic worms are among the organism suitable for mounting with CMCP. The CMCP mounting provide then with rapid relaxation, death, and clearing action.
Catalog No. 50-190-0430
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.