Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNQX disodium salt, Tocris Bioscience™

Catalog No. p-4783368 Shop All R&D Systems Products
Click to view available options
Quantity:
1 mg
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent non-NMDA iGluR antagonist; more water soluble form of CNQX (Cat. No. 0190)

More water soluble disodium salt of the AMPA/kainate antagonist CNQX (Cat. No. 0190).

Chemical Identifiers

CAS 479347-85-8
Molecular Formula C9H2N4Na2O4
Molecular Weight (g/mol) 276.119
InChI Key YCXDDPGRZKUGDG-UHFFFAOYSA-L
Synonym cnqx disodium salt, disodium 6-cyano-7-nitroquinoxaline-2,3-diolate, 6-cyano-7-nitroquinoxaline-2,3-dione disodium, sodium 6-cyano-7-nitroquinoxaline-2,3-bis olate, cnqx disodium salt hydrate, 6-quinoxalinecarbonitrile,1,2,3,4-tetrahydro-7-nitro-2,3-dioxo, 6-cyano-7-nitroquinoxaline-2,3-dione disodium salt hydrate, 6-cyano-7-nitroquinoxaline-2,3-dione 2na, 1,2,3,4-tetrahydro-7-nitro-2,3-dioxoquinoxaline-6-carbonitrile disodium
PubChem CID 6093155
IUPAC Name disodium;6-cyano-7-nitroquinoxaline-2,3-diolate
SMILES C1=C(C(=CC2=C1N=C(C(=N2)[O-])[O-])[N+](=O)[O-])C#N.[Na+].[Na+]

Specifications

Quantity 1 mg
Formula Weight 276.12
Percent Purity >99%
Chemical Name or Material CNQX disodium salt
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.