Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNQX, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
CNQX is an AMPA and kainate receptor antagonist (IC50 values are 0.3 μM and 1.5 μM for AMPA and kainate receptors, respectively). CNQX is also an antagonist at the glycine modulatory site on the NMDA receptor complex (IC50 = 25 μM). CNQX can be used to isolate GABAA receptor-mediated spontaneous inhibitory postsynaptic currents and antagonizes non-NMDA receptor-mediated responses in cultured cerebellar granule cells. CNQX shows neuroprotective effects in models of ischemia and inhibits seizure-like activity in hippocampal neurons.
CNQX Disodium Salt (Cat. No. 1045) also available.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 232.16 |
Percent Purity | >99% |
Chemical Name or Material | CNQX |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction