Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ CNV Dopamine

Catalog No. 63545 Shop All R&D Systems Products
Click to view available options
:
5 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Caged dopamine; photolysed by UV light

CNV Dopamine is a carboxynitroveratryl (CNV)-caged dopamine (Cat. No. 3548); releases dopamine upon ultraviolet light stimulation (365 nm). Induces dopaminergic currents upon photolysis in brain slices expressing wild-type or GFP-labeled D2 receptors. Displays no decrease in current (rundown) with UV light flash duration of up to 100 ms. Pharmacologically inactive prior to photolysis.

Specifications

Quantity 5 mg
Target Caged Compounds
Molecular Formula C18H20N2O8
Formula Weight 2879
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.