Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Cobalt(ous) Chloride, Hexahydrate
SCI_ED

Catalog No. p-4416365 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
Packaging:
Lab Grade
Reagent Grade
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7791-13-1, 7732-18-5
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
Synonym Cobalt(II) Chloride Hexahydrate, Cobalt muriate hexahydrate
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Color Purple
Physical Form Solid
Quantity 100 g
Assay Percent Range <75%w/v
Linear Formula CoCl2 · 6H2O
Formula Weight 237.95
Grade Reagent
Packaging Lab Grade
Density 1.924
Chemical Name or Material Cobalt(ous) Chloride, Hexahydrate
ShelfLife 2 years
DOTInformation DOT Class 8, III, : Corrosive
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.