Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Cobalt(ous) Nitrate, Hexahydrate
SCI_ED

Catalog No. p-4416366 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade
Reagent Grade

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 10026-22-9
Molecular Formula CoH12N2O12
Molecular Weight (g/mol) 291.03
MDL Number MFCD00149647
InChI Key QGUAJWGNOXCYJF-UHFFFAOYSA-N
Synonym cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate
PubChem CID 24821
ChEBI CHEBI:86214
IUPAC Name λ²-cobalt(2+) hexahydrate dinitrate
SMILES O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Red
Physical Form Solid
Quantity 100 g
Assay Percent Range >98%
Linear Formula Co(NO3)2 · 6H2O
Formula Weight 291.05
Packaging Lab Grade
Density 1.88g/mL
Chemical Name or Material Cobalt(ous) Nitrate, Hexahydrate
ShelfLife 1 year
DOTInformation DOT Class 5.1, II, : Oxidizer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.