Learn More
Hach Company COD Standard Solution, 300 mg/L as COD (NIST), 200 mL
Supplier:  Hach Company 1218629
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
For accuracy checks in Chemical Oxygen Demand (COD) determinations. 200 mL bottle
Catalog No. 50-199-4854