Learn More
Hach Company COD Digestion Vials, Manganese III, pk/150
Supplier: Hach Company 2623415
For determination of Chemical Oxygen Demand (COD) by the Manganese III Reactor Digestion Method. Hach Method 10067 Range: 30 - 1000 mg/L COD Pack of 150 vials.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.