Learn More
Hach Company COD Digestion Vials, Manganese III, pk/25
Supplier:  Hach Company 2623425
Encompass_Preferred
For determination of Chemical Oxygen Demand (COD) by the Manganese III Reactor Digestion Method. Hach Method 10067 Range: 30 - 1000 mg/L COD Pack of 25 vials.
Catalog No. 50-199-6384
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.