Learn More
Hach Company Chemical Oxygen Demand (COD) TNTplus Vial Test, ULR (1-60 mg/L COD), 25 Tests
Supplier:  Hach Company TNT820
Encompass_Preferred
Hach TNTplus COD vial test reagents are incredibly simple to use and cut down significantly on waste generation compared to other COD test kits. TNT820 is based on Hach Method 10211 for virtually any type of water sample. Hach TNTplus COD vial tests are available in multiple ranges. TNT820 provides a result in the Ultra Low Range of 1 - 60 mg/L COD and is often used for Chemical Oxygen Demand measurement in wastewater effluent. Hach TNT820 does not require a separate blanking vial for further waste reduction. Make the switch to TNTplus COD reagents today!For determination of Chemical Oxygen Demand (COD) by the Reactor Digestion Method. Simply pipette in sample, digest, and read in your Hach Spectrophotometer. 25 tests per box.
Catalog No. 50-199-8111
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.