Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ COL1A1 Split FISH Probe

Catalog No. 89940098 Shop All Abnova Corporation Products
Click to view available options
Quantity:

Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.

Specifications

For Use With (Application)

FISH-Ce

Includes

DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe

Quantity

100 μL

Content And Storage

Store at 4°C in the dark.

Description

DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe

Gene ID (Entrez)

1277

Format

Liquid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.