Learn More
Electron Microscopy Sciences Colloidal Alumina, 1 Pint 16 OZ
Supplier: Electron Microscopy Sciences 5036505
As an acidic colloidal dispersion, Colloidal Alumina chemical-mechanical action provides superior surfaces on germanium, silicon, gallium arsenide, zinc sulphide, fluorides, sapphire, ferrite's and silicon carbide. Low scatter finishes can be obtained on pure metals such as nickel, tungsten, copper, beryllium, steel and aluminum. Epoxy resin materials and plastics can also be fully polished using this colloidal dispersion.
Colloidal Alumina can be used full strength or diluted. Polyurethane and short nap cloths are recommended.
Properties: pH: 3 - 3.5; solids: 20%; viscosity: thixotropic.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.